Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GST Rabbit Polyclonal Antibody

Supplier: BioVision

3997-100
10005-200EA 252.35 USD InStock
10005-200
Anti-GST Rabbit Polyclonal Antibody
Antibodies
Glutathione S-transferase (GST) is a widely used protein fusion partner, since it provides an easily detectable Tag and also a simple purification process with little effect on the biological function of the protein of interest.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml, frozen & paraffin

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty