Anti-RFP Rabbit Polyclonal Antibody

Supplier: BioVision
AB62341-100
10005-188EA 459.05 USD
10005-188
Anti-RFP Rabbit Polyclonal Antibody
Antibodies
Since the molecular cloning of GFP cDNA and demonstration of GFP as a functional transgene, GFP has become a powerful tool with exciting applications in developmental, cell and molecular biology. RFP is the recent discovered protein that has the similar function and applications as GFP. Either GFP or RFP fluorescence is not species specific and can be expressed in bacteria, yeast, plant and mammalian cells. GFP or RFP can fuse with proteins of interest without interfering significantly with their assembly and function.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR