Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S-Tag Rabbit Polyclonal Antibody

Supplier: BioVision

3990-100
10005-180EA 252.35 USD InStock
10005-180
Anti-S-Tag Rabbit Polyclonal Antibody
Antibodies
S-tag is an epitope tag composed of a 15 residue peptide derived from pancreatic ribonuclease A

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: S-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S-Tag And S-Tag Fusion Proteins

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty