Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-D-Tag Rabbit Polyclonal Antibody

Supplier: BioVision

3989-100
10005-178EA 272.95 USD InStock
10005-178
Anti-D-Tag Rabbit Polyclonal Antibody
Antibodies
DYKDDDDK Tag antibody detects over-expressed proteins containing the D epitope tag fused to either the amino- or carboxy-terminus of targeted proteins. Epitope tags are usually for the labeling and detection of proteins using immunoblotting, immunoprecipitation and immunostaining techniques. Due to their small size, they are unlikely to affect the tagged protein’s biochemical properties. The DYKDDDK epitope tag is widely used to detect expression of recombinant proteins in bacteria, yeast, insect and mammalian systems

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: D-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty