Anti-T7 Epitope -Tag Rabbit Polyclonal Antibody

Supplier: BioVision
3987-100
10005-174EA 309.39 USD
10005-174
Anti-T7 Epitope -Tag Rabbit Polyclonal Antibody
The T7-tag is an 11 amino acid sequence encoded in the leader sequence of T7 bacteriophage gene10. T7 is used in many expression vectors including the pET system. Hence, the anti-T7-tag antibody is useful for studying expression of recombinant T7-tagged proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: T7 Epitope -Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR