Anti-SBP-Tag Rabbit Polyclonal Antibody

Supplier: BioVision
3986-100
10005-172EA 356.1 USD
10005-172
Anti-SBP-Tag Rabbit Polyclonal Antibody
Streptavidin Binding Peptide (SBP) tag is very versatile and can be inserted into expression vectors for easy detection of recombinant proteins and purification by use of affinity columns. The full sequence for SBP-tag is 38 amino acids long and exhibits a nanomolar scale binding affinity for streptavidin.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SBP-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR