Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-V5 Tag Rabbit Polyclonal Antibody

Supplier: BioVision

3985-100
10005-170EA 272.95 USD InStock
10005-170
Anti-V5 Tag Rabbit Polyclonal Antibody
Antibodies
V5-tag is a 14 amino acid peptide derived from the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The V5-tagged protein expression system is preferably used in various laboratories. Hence, the anti-V5-Tag antibody is a useful tool for monitoring of the V5-tagged protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty