Anti-SREBF2 Rabbit Polyclonal Antibody

Supplier: BioVision
3961-100
10005-154EA 303.55 USD
10005-154
Anti-SREBF2 Rabbit Polyclonal Antibody
SREBP (Sterol Regulatory Element Binding Protein) 1 and 2 are transcription factors th at regulates cholesterol homeostasis. SREBP proteins are attached to the endoplasmic reticulum and nuclear envelope. SREBP proteins undergo proteolytic cleavage in response to conditions of low cellular sterol. Upon activation of SREBP 1, a soluble NH2 cleavage fragment translocate to the nucleus and activates transcription of enzymes and other proteins required for cholesterol synthesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SREBF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR