Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CBP-Tag Rabbit Polyclonal Antibody

Supplier: BioVision

3960-100
10005-150EA 288.4 USD InStock
10005-150
Anti-CBP-Tag Rabbit Polyclonal Antibody
Antibodies
Calmodulin binding peptide (CBP) tag is a fusion protein that is useful for immunoaffinity purification of proteins expressed in E.coli. The CBP tag binds to calmodulin resin in the presence of low concentration of calcium and can be eluted with a neutral buffer. The antibody is suitable for identifying CBP-fusion proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CBP-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty