Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PI3 kinase Rabbit Polyclonal Antibody

Supplier: BioVision

3959-100
10005-146EA 252.35 USD InStock
10005-146
Anti-PI3 kinase Rabbit Polyclonal Antibody
Antibodies
Phosphatidylinositol 3-kinase (PI 3-kinase), is a phosphoinositide-specific protein kinase that plays a central role in mitogenic signal transduction leading to cellular regulation, cell growth and differentiation, and oncogenesis. PI 3-kinase associates with, and is activated by a wide range of tyrosine kinase receptors including EGF, PDGF, CSF-1, erbB-3 and insulin receptor, non-receptor protein-tyrosine kinases of the src family (e.g. pp60src), and crk and abl proto-oncogene products. PI 3-kinase is involved in signal transduction in terminally differentiated, activated cells like platelets and neutrophils.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: PI3 kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty