Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S100 Rabbit Polyclonal Antibody

Supplier: BioVision

3958-100
10005-142EA 252.35 USD InStock
10005-142
Anti-S100 Rabbit Polyclonal Antibody
Antibodies
S-100 protein is expressed from the beginning of lipidogenesis and may act as a factor regulating lipid storage and body fat formation. It can be used as a reliable biochemical marker of human fat cell differentiation and for distinguishing them from the mesenchymal and the fibroblast cells in the human embryonal subcutis.

Recommended Dilutions: Western Blot: 1–4 µg/ml).

Type: Primary
Antigen: S100
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty