Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE3B Rabbit Polyclonal Antibody

Supplier: BioVision

3956-200
10005-134EA 267.72 USD InStock
10005-134
Anti-PDE3B Rabbit Polyclonal Antibody
Antibodies
The phosphodiesterase type-3 (PDE3) family is comprised of 2 isoforms, PDE3A and PDE3B.
Phosphodiesterases (PDE) are crucial in the regulation of the intracellular level of the second messenger cAMP by hydrolyzing cAMP to 5’AMP. PDE3B has two splice variants that are expressed in adipocyte tissue, hepatocyte, kidney, spermatocytes and embryonic neuroepithelium. Both PDE3A and PDE3B are potential targets for the treatment of obesity.

Recommended Dilutions: Western Blot: 1–4 µg/ml).

Type: Primary
Antigen: PDE3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty