Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AS160/TBC1 Rabbit Polyclonal Antibody

Supplier: BioVision

3954-100
10006-002EA 252.35 USD InStock
10006-002
Anti-AS160/TBC1 Rabbit Polyclonal Antibody
Antibodies
AS160 is a 160 kDa Akt serine/threonine kinase substrate that plays a critical role in regulating insulin-stimulated exocytosis of glucose transporter GLUT4. Increased in insulin treatment stimulate phophorylation of AS160 that regulates GLUT4 translocation via inactivation of the Rab GTPase function.

Recommended Dilutions: Western Blot: 1–4 µg/ml).

Type: Primary
Antigen: AS160/TBC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty