Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMGCR Rabbit Polyclonal Antibody

Supplier: BioVision

3952-100
10005-996EA 288.4 USD InStock
10005-996
Anti-HMGCR Rabbit Polyclonal Antibody
Antibodies
HMG-CoA reductase (HMGCR) is the rate-limiting enzyme for cholesterol biosynthesis. HMGCR is regulated via a negative feedback mechanism mediated by sterols and non-sterol metabolites derived from the product of the reductase reaction. Statin, a class of cholesterol-lowering drμgs inhibits HMGCR by lowering plasma cholesterol.

Recommended Dilutions: Western Blot: 0.5–4 µg/ml).

Type: Primary
Antigen: HMGCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty