Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STK11 Rabbit Polyclonal Antibody

Supplier: BioVision

3950-100
10005-988EA 252.35 USD InStock
10005-988
Anti-STK11 Rabbit Polyclonal Antibody
Antibodies
LKB1 is a tumor suppressor that belongs to the serine/threonine kinase family. Mutation in LKB1 is link to Peutz Jeghers syndrome (PJS), a disorder characterized by the growth of polyps in the gastrointestinal tract. LKB1 regulate AMPK signaling pathway and may also play a role in chromatin remodeling, cell cycle arrest and cell polarity

Recommended Dilutions: Western Blot: 0.5–4 µg/ml).

Type: Primary
Antigen: STK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty