Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PEPCK-C Rabbit Polyclonal Antibody

Supplier: BioVision

3949-100
10005-984EA 252.35 USD InStock
10005-984
Anti-PEPCK-C Rabbit Polyclonal Antibody
Antibodies
Phosphoenolpyruvate carboxykinase (PEPCK) is a key enzyme in stimulating glucose production. PEPCK catalyzes the conversion of oxaloacetate to phosphoenolpyruvate. There are two forms of PEPCK.,the cytosolic form also known as PEPCK-C and the mitochondrial form, or PEPCK-M.

Recommended Dilutions: Western Blot: 0.5–4 µg/ml).

Type: Primary
Antigen: PEPCK-C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty