Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LPL Rabbit Polyclonal Antibody

Supplier: BioVision

3947-100
10005-976EA 262.65 USD InStock
10005-976
Anti-LPL Rabbit Polyclonal Antibody
Antibodies
Lipoprotein lipase (LPL) is mainly expressed in the heart, muscle and adipose tissue. LPL is a homodimer that mediates the hydrolysis of triglycerides of very low density lipoproteins and also function as ligand/bridging factor for receptor-mediated lipoprotein uptake. Defects in LPL may result in hypertriglceridemia.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty