Anti-GLUT4 Rabbit Polyclonal Antibody

Supplier: BioVision
3945-200
10004-950EA 286.04 USD
10004-950
Anti-GLUT4 Rabbit Polyclonal Antibody
Insulin stimulates glucose uptake in the muscles and adipose tissues by activating GLUT4 (Glucose Transporter Type 4). GLUT 4 is a transmembrane protein that facilitates glucose transport from storage sites to cell surface. Human insulin resistance involves a defect in GLUT4 that leads to accumulation in a dense membrane compartment from which insulin is unable to recruit GLUT4 to the cell surface.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: GLUT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR