Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pyruvate Dehydrogenase Phosphatase 2 Rabbit Polyclonal Antibody

Supplier: BioVision

3944-200
10004-946EA 252.35 USD InStock
10004-946
Anti-Pyruvate Dehydrogenase Phosphatase 2 Rabbit Polyclonal Antibody
Antibodies
PDP2, Pyruvate dehydrogenase phosphatase 2, catalyzes the dephosphorylation and activation of E1 component to reverse the effects of pyruvate dehydrogenase kinases. Pyruvate dehydrogenase (E1) is one of the three components (E1, E2, and E3) of the large pyruvate dehydrogenase complex. 

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Pyruvate Dehydrogenase Phosphatase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty