Anti-SUV39H1 Rabbit Polyclonal Antibody

Supplier: BioVision
3942-100
10005-098EA 286.04 USD
10005-098
Anti-SUV39H1 Rabbit Polyclonal Antibody
The histone methyltransferase KMT1A / SUV39H1 is the mammalian homologue of the Drosophila PEV modifier Su(var)3-9 protein. SUV39H1 contains the SET and chromo domains. In eukaryotic cells, SUV39H1 and the methyl-lysine binding protein HP1 interact to repress transcription at heterochromatic sites. SUV39H1 methylates histone H3 on lysine 9, which generates a binding site for HP1 proteins that plays a role in heterochromatin formation and gene silencing.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: SUV39H1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR