Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TP2 Rabbit Polyclonal Antibody

Supplier: BioVision

3941-100
10005-094EA 252.35 USD InStock
10005-094
Anti-TP2 Rabbit Polyclonal Antibody
Antibodies
Telomerase is an RNA-dependant DNA polymerase that catalyzes the addition of telemoric repeat, TTAGGG to chromosome ends. Telomerase is composed of an internal telomerase RNA template (TERC) and the enzyme, telomerase reverse transcriptase (TERT or TP2). Telomerase expression is usually repressed in postnatal somatic cells resulting in shortening of telomeres. Overexpression of telomerase may be implicated with oncogenesis.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: TP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty