Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADRB3 Rabbit Polyclonal Antibody

Supplier: BioVision

3939-200
10005-088EA 272.95 USD InStock
10005-088
Anti-ADRB3 Rabbit Polyclonal Antibody
Antibodies
The neurotransmitter/hormone adrenaline (epinephrine, adrenalin) plays a central role in the mammalian stress response, increasing heart rate, raising blood pressure, and increasing blood glucose levels. Adrenaline activates both alpha-adrenergic receptors and beta-adrenergic receptors. There are mainly three subtypes of beta adrenergic receptors known, beta1, beta2, and beta3. These receptors are expressed primarily in the heart, respiratory tissue, and adipose tissue. Beta3-receptors are abundant in brown adipocytes and play vital roles in lipolysis and thermoregulation. Beta adrenergic receptors are gaining much interest among researches in the studies of type-2 diabetes mellitus and obesity. It is also being considered as a therapeutic target for heart failure.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty