Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CFD Rabbit Polyclonal Antibody

Supplier: BioVision

3937-100
10005-080EA 252.35 USD InStock
10005-080
Anti-CFD Rabbit Polyclonal Antibody
Antibodies
Adipsin is a serine protease secreted by adipose tissue, and it has also been found in monocytes/macrophages. Adipsin has been identified to be similar to complement Factor D. Adipsin cleaved complement Factor B when the later form complexed with activated complement component C3. Low level expression of Adipsin has been linked to obesity in mouse model sμggesting that defects in Adipsin is the result of this systemic disorder.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CFD
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty