Anti-PRMT5 Rabbit Polyclonal Antibody

Supplier: BioVision
3935-100
10004-936EA 286.04 USD
10004-936
Anti-PRMT5 Rabbit Polyclonal Antibody
Protein arginine N-methyltransferase 5 (PRMT-5) is a member of the PRMT family that localizes to the cytoplasm in many types of tissues. At least three types of PRMT enzymes have been identified that shows essential regulatory functions by methylation of key proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PRMT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR