Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNA Binding Protein-7 Rabbit Polyclonal Antibody

Supplier: BioVision

3933-100
10005-074EA 252.35 USD InStock
10005-074
Anti-DNA Binding Protein-7 Rabbit Polyclonal Antibody
Antibodies
DNA binding protein 7 (DBP-7) is also known as Chd7. Mutations in the Chd7 gene is believed to be a major implication of CHARGE syndrome. CHARGE syndrome is associated with anomalies such as malformations of the heart, inner ear, retina and cranial nerve defects.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DNA Binding Protein-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty