Anti-TLR10 Rabbit Polyclonal Antibody

Supplier: BioVision
3932-100
10004-934EA 286.04 USD
10004-934
Anti-TLR10 Rabbit Polyclonal Antibody
The Toll-like receptor (TLR) family of proteins are characterized by a highly conserved Toll homology (TH) domain, which is essential for Toll-induced signal transduction. TLRs require adapter molecule such as MyD88 and TIRAP to activate various kinases and transcription factors. TLR10 is homologous to TLRs 1 and 6 and is highly expressed in the lymphoid tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TLR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR