Anti-TLR11 Rabbit Polyclonal Antibody

Supplier: BioVision
3931-100
10004-930EA 286.04 USD
10004-930
Anti-TLR11 Rabbit Polyclonal Antibody
The Toll-like receptor (TLR) family of proteins are characterized by a highly conserved Toll homology (TH) domain, which is essential for Toll-induced signal transduction. TLRs require adapter molecule such as MyD88 and TIRAP to activate various kinases and transcription factors.TLR11 is activated by uropathogenic bacteria and may play a role in preventing urogenital infections.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TLR11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR