Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMI1 Rabbit Polyclonal Antibody

Supplier: BioVision

3930-100
10004-926EA 252.35 USD InStock
10004-926
Anti-BMI1 Rabbit Polyclonal Antibody
Antibodies
Polycomb group (PcG) genes encode chromatin proteins that are involved in the maintenance of cellular memory throμgh epigenetic chromatin modifications. Bmi-1 has been identified among the PcG genes as a potent oncogene that plays an important role in the regulation of cell proliferation by suppressing the p16-dependant pathway. Bmi-1 may be use as a therapeutic target for studying stem cell proliferation and renewal.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Dog, Cat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty