Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PINK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3929-100
10004-922EA 262.65 USD InStock
10004-922
Anti-PINK1 Rabbit Polyclonal Antibody
Antibodies
PINK1 (PTEN-induced putative kinase 1) is a serine/threonine protein kinase found in mitochondria. PINK1 protects cell from .stress-induced mitochondrial dysfunction. Mutation in this gene is associated with the early onset of Parkinson's disease.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty