Anti-ICAM2 Rabbit Polyclonal Antibody

Supplier: BioVision
3926-100
10004-910EA 286.04 USD
10004-910
Anti-ICAM2 Rabbit Polyclonal Antibody
Cell adhesion molecules (CAMs) are a family of closely related cell surface glycoproteins involved in cell-cell interactions during growth and are thoμght to play an important role in embryogenesis and development. The family contains NCAM (for neuronal cell adhesion molecule), ICAM (for intercellular adhesion molecule), VCAM (vascular cell adhesion molecule), PECAM-1 (platelet/endothelial cell adhesion molecule-1), etc. ICAM-2 is involved in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: ICAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR