Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Influenza A Virus Hemagglutinin Rabbit Polyclonal Antibody

Supplier: BioVision

3924-100
10004-904EA 252.35 USD InStock
10004-904
Anti-Influenza A Virus Hemagglutinin Rabbit Polyclonal Antibody
Antibodies
Influenza A viruses are surface glycoprotein, single stranded, segmented RNA viruses that are found in birds. Influenza A subtypes are based on the virus glycoproteins hemagglutinin
(HA) and neuraminidase (NA). There are 16 different HA antigens and nine different N antigens for influenza A virus that have been isolated from wild birds.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Influenza A Virus Hemagglutinin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty