Anti-EMP3 Rabbit Polyclonal Antibody

Supplier: BioVision
3923-100
10004-900EA 286.04 USD
10004-900
Anti-EMP3 Rabbit Polyclonal Antibody
Epithelial membrane protein 3 (EMP3) is a multi pass membrane protein that has two N- glycosylation and four transmembrane domain. EMP3 is a myelin-related protein involves in cell-cell interactions and cell proliferation. Over-expression of EMP3 may indicate carcinogenesis or the development of oligodendrogial tumors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: EMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR