Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Serpin A6 Rabbit Polyclonal Antibody

Supplier: BioVision

3906-100
10004-894EA 252.35 USD InStock
10004-894
Anti-Serpin A6 Rabbit Polyclonal Antibody
Antibodies
Serpin A6 is a secreted glycoprotein that belongs to the serpin family of protease inhibitors.
Serpin A6 is produced from hepatocytes in the liver and is involved in the transport of glucocorticoids and progestins in the blood.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Serpin A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty