Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LCN2 Rabbit Polyclonal Antibody

Supplier: BioVision

3902-100
10004-888EA 252.35 USD InStock
10004-888
Anti-LCN2 Rabbit Polyclonal Antibody
Antibodies
Lipocalin 2 is from a family of proteins that are involved in the transportation of small hydrophobic molecules including steroids, retinoids, bilin and retinods. Lipocalins have been linked to many biolchemical processes such as immune response, pheromone transport, biological prostaglandin synthesis, retinoid binding, and cancer cell interactions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty