Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGI-58 Rabbit Polyclonal Antibody

Supplier: BioVision

3901-100
10004-884EA 252.35 USD InStock
10004-884
Anti-CGI-58 Rabbit Polyclonal Antibody
Antibodies
Comparative Gene Identification-58 (CGI-58) is a causal gene of Dorfman-Chanarin syndrome, a neutral lipid storage disease characterized by the presence of intracellular lipid droplets in tissues. CGI-58 comes from a large family of proteins characterized by an α/β hydrolase fold that activates adipose triglyceride lipase thus, may involve in lipid metabolism.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CGI-58
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty