Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMP2 Rabbit Polyclonal Antibody

Supplier: BioVision

3900-100
10004-880EA 262.65 USD InStock
10004-880
Anti-LAMP2 Rabbit Polyclonal Antibody
Antibodies
Lysosomal-associated membrane protein 2 (LAMP2) also known as CD107B (Cluster of Differentiation 107B) is a membrane glycoprotein. LAMP2 plays an important role in the normal function of lysosome which includes protecting lysosomal membrane from autodigestion, maintaining lysosome acidity, and lysosome cellular adhesion.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty