Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HPX Rabbit Polyclonal Antibody

Supplier: BioVision

3899-200
10005-020EA 252.35 USD InStock
10005-020
Anti-HPX Rabbit Polyclonal Antibody
Antibodies
Hemopexin (HPX) is a plasma protein that has the highest binding affinity for heme. HPX prevents heme-mediated oxidative stress and heme-bound iron loss by transporting heme to the liver for breakdown.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HPX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty