Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NFkB p105 Rabbit Polyclonal Antibody

Supplier: BioVision

3897-200
10005-958EA 252.35 USD InStock
10005-958
Anti-NFkB p105 Rabbit Polyclonal Antibody
Antibodies
NFkB was identified as a sequence specific transcriptional activator that binds to the intronic enhancer of kappa light chain gene in B lymphocytes. NFkB is activated by a wide variety of stimuli such as lipopolysaccharide (LPS), pro-inflammatory cytokines, and viral infection. The active NFkB heterodimer is translocated into the nucleus and induces gene expression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NFkB p105
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty