Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLAME Rabbit Polyclonal Antibody

Supplier: BioVision

3895-100
10005-950EA 252.35 USD InStock
10005-950
Anti-BLAME Rabbit Polyclonal Antibody
Antibodies
BLAME (B Lymphocyte Activator Macrophage Expressed) also called SLAMF8. BLAME is a type I transmembrane protein that belongs to the CD2 family of Ig superfamily receptors. BLAME is mostly expressed on macrophages and dendritic cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BLAME
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty