Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DKK4 Rabbit Polyclonal Antibody

Supplier: BioVision

3894-100
10005-016EA 252.35 USD InStock
10005-016
Anti-DKK4 Rabbit Polyclonal Antibody
Antibodies
Xenopus Dickkopf (Dkk) was initially discovered as a Wnt antagonist that plays an important role in head formation. By far, four members of Dkk have been identified in mammals. Each Dkk molecule contains two conserved cysteine-rich domains which are separated by a 50-55 amino acid linker region. Dkk4 is a secreted glycoprotein and has three immune-reactive sites at 15-17kDa, 30-32kDa and at 40kDa.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DKK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty