Anti-FOXH1 Rabbit Polyclonal Antibody

Supplier: BioVision
3892-100
10004-872EA 286.04 USD
10004-872
Anti-FOXH1 Rabbit Polyclonal Antibody
FoxH1 (also known as Forkhead box H1 or FAST-1) is a transcription factor that shares close homology with Xenopus FAST-1. FoxH1 binds to SMAD2/4 and activates activin response throμgh a DNA binding motif region that consist of TGT(G/T)(T/G)ATT sequence. FoxH1 binding to Smad2/Smad4/FAST1 forms a transcriptionally complex structure that can induced TGFβ signal transduction.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FoxH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR