Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF4 Rabbit Polyclonal Antibody

Supplier: BioVision

3890-100
10005-008EA 252.35 USD InStock
10005-008
Anti-TNFRSF4 Rabbit Polyclonal Antibody
Antibodies
OX40 is also known as CD134 is a member of the tumor necrosis factor receptor superfamily (TNFR). OX40 plays an important role in CD4+ and CD8+T-cells and B-cells response. Binding of OX40 to its ligand, OX40L provides co-stimulatory signal to effector T-cells without CD28 activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNFRSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty