Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BORIS Rabbit Polyclonal Antibody

Supplier: BioVision

3888-100
10005-000EA 262.65 USD InStock
10005-000
Anti-BORIS Rabbit Polyclonal Antibody
Antibodies
Brother of the regulator of imprinted sites (BORIS) is a transcription factor also known as CCCTC- binding factor like protein (CTCF). Boris is usually expressed at a high level in the testis which correlates with male germ cell development and is also abnormally activated in a broad range of human cancers. Boris can be found in the nucles as well as in the cytoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BORIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty