Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SP3 Rabbit Polyclonal Antibody

Supplier: BioVision

3887-100
10004-996EA 252.35 USD InStock
10004-996
Anti-SP3 Rabbit Polyclonal Antibody
Antibodies
Sp transcription factor family has a highly conserved DNA-binding domain composed of zinc fingers. The Sp family is comprised of Sp1, Sp2, Sp3 and Sp4 that regulates transcription by binding to consensus GC- and GT-box regulatory elements in the target genes. Sp1, Sp3 and Sp4 have a high binding capacity for GC box while Sp2 binds weakly to the GT box. Sp3 has been shown to have to a bifunctional transcription factor that can either stimulates or represses the transcription of various genes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty