Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3883-100
10004-862EA 252.35 USD InStock
10004-862
Anti-SPK1 Rabbit Polyclonal Antibody
Antibodies
Sphingosine Kinase (SphK) is a conserved lipid kinase that catalyzes the phosphorylation of sphingolipid sphingosine to sphingosine-1-phosphate (SIP). SIP receptors coupled to cell surface G protein to regulate cell growth, survival, motility and inflammatory responses. There are two types of SphK, SphK1 and SphK2. SphK1 is found in the cytosol and migrates to the membrane when activated. SphK1 has been associated with cell growth, prevention of apoptosis and cellular transformation. SphK2 is found mainly in the nucleus and in contrast to SphK1, it enhances the rate of apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty