Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MFN2 Rabbit Polyclonal Antibody

Supplier: BioVision

3882-100
10004-858EA 252.35 USD InStock
10004-858
Anti-MFN2 Rabbit Polyclonal Antibody
Antibodies
Mitofusin 2 (Mfn 2) is mostly expressed in the heart and muscle tissues. It is a transmembrane protein that mediates mitochondria fusion and plays a central role in the maintenance of mitochondrial morphology. A GTPase domain is required for the function of Mitofusin proteins. Mutations in Mfn2 can lead to Charcot-Marie-Tooth disease, a common inherited disorder of the peripheral nervous system. Mfn2 may also be associated with obesity and/or apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MFN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty