Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOTCH1 Rabbit Polyclonal Antibody

Supplier: BioVision

3881-100
10004-856EA 252.35 USD InStock
10004-856
Anti-NOTCH1 Rabbit Polyclonal Antibody
Antibodies
Notch proteins are transmembrane receptor that regluate cell fate decisions. Four Notch homologs have been identified in mammals namely Notch 1, Notch 2, Notch 3 and Notch 4. The membrane ligand for Notch includes Jagged 1, Jagged 2 and Delta. Notch 1 is a 270 kDa transmembrane receptor that can be activated by Delta (Dl) ligand. Activation of Notch 1 will result in the proteolytic cleavage of Notch 1 into a 120 kDa length protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty