Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMC4 Rabbit Polyclonal Antibody

Supplier: BioVision

3876-100
10004-846EA 267.72 USD InStock
10004-846
Anti-SMC4 Rabbit Polyclonal Antibody
Antibodies
The SMC (structural maintenance of chromosomes) family of proteins form heterodimeric complexes that modulate sister chromatid cohesion and chromosome condensation for mitosis. The two distinct classes of SMC protein complexes are comprised of SMC1 (also designated SB1.8) with SMC3 (also designated hCAP and Bamacan), and SMC2 (also designated hCAP-E for chromosome-associated polypeptide E) with hCAP-C. The SMC2/hCAP-C complex is required for mitotic chromosome condensation and functions independently of the SMC1/SMC3 complex during the cell cycle. Both SMC2 and hCAP-C are expressed throughout the cell cycle in a variety of cell lines, including HeLa cells, Daudi B cells, Jurkat T cells, SK2 neuronal cells, and HepG2 liver cells.

Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: SMC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty