Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCD Rabbit Polyclonal Antibody

Supplier: BioVision

3875-100
10004-842EA 252.35 USD InStock
10004-842
Anti-SCD Rabbit Polyclonal Antibody
Antibodies
Stearoyl-CoA desaturase (SCD) is a regulatory enzyme involved in the synthesis of oleate and palmitoleate. Oleate and palmitoleate are the main components of membrane phospholipid, triglycerides and cholesterol esters. SCD plays an important role in cellular cholesterol homeostasis and also in the secretion processes of triacylglycerol and phospholipid. Inhibition of SCD could be a useful treatment for metabollic disorders such as obesity and hepatic steatosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty