Anti-FURIN Rabbit Polyclonal Antibody

Supplier: BioVision
3873-100
10004-834EA 286.04 USD
10004-834
Anti-FURIN Rabbit Polyclonal Antibody
Furin is a ubiquitous protein located within the trans-Golgi network. It is found in many tissues and cell lines including MDCK, HeLa, HepG2 and NIH-3T3. Furin is a proprotein convertase that is responsible for the proteolytic maturation of many precursor proteins that are secreted from the constitutive secretory pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Furin/PACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR